Anti-ATG3 Mouse Monoclonal Antibody [clone: OTI3H2]

Supplier: OriGene
TA503346
ORIGTA503346EA 621 EUR
ORIGTA503346
Anti-ATG3 Mouse Monoclonal Antibody [clone: OTI3H2]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ATG3 (autophagy related 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR