Anti-CCM2 Mouse Monoclonal Antibody [clone: OTI2F9]

Supplier: OriGene
ORIGTA503414EA 603 EUR
ORIGTA503414
Anti-CCM2 Mouse Monoclonal Antibody [clone: OTI2F9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: CCM2 (cerebral cavernous malformation 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR