Anti-CTBP1-AS2 Mouse Monoclonal Antibody [clone: OTI2C6]

Supplier: OriGene
ORIGTA503479EA 603 EUR
ORIGTA503479
Anti-CTBP1-AS2 Mouse Monoclonal Antibody [clone: OTI2C6]
Antibodies
Recommended Dilutions: Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CTBP1-AS2 (CTBP1 antisense RNA 2 (head to head))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR