Anti-LGALS3BP Mouse Monoclonal Antibody [clone: OTI3D6]

Supplier: OriGene
ORIGTA503500EA 603 EUR
ORIGTA503500
Anti-LGALS3BP Mouse Monoclonal Antibody [clone: OTI3D6]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LGALS3BP (lectin, galactoside-binding, soluble, 3 binding protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR