Anti-ERBB2 Mouse Monoclonal Antibody [clone: OTI12G4]

Supplier: OriGene
ORIGTA503527EA 603 EUR
ORIGTA503527
Anti-ERBB2 Mouse Monoclonal Antibody [clone: OTI12G4]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: ERBB2 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR