Anti-MTFMT Mouse Monoclonal Antibody [clone: OTI1C1]

Supplier: OriGene
TA503547
ORIGTA503547EA 621 EUR
ORIGTA503547
Anti-MTFMT Mouse Monoclonal Antibody [clone: OTI1C1]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: MTFMT (mitochondrial methionyl-tRNA formyltransferase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR