Anti-CDKN3 Mouse Monoclonal Antibody [clone: OTI4D8]

Supplier: OriGene
ORIGTA503592EA 616 EUR
ORIGTA503592
Anti-CDKN3 Mouse Monoclonal Antibody [clone: OTI4D8]
Antibodies
Recommended Dilutions: Western Blot: 1:200-500; Immunofluorescence: 1:100

Type: Primary
Antigen: CDKN3 (cyclin-dependent kinase inhibitor 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR