Anti-ACSS2 Mouse Monoclonal Antibody [clone: OTI2C6]

Supplier: OriGene
TA503610
ORIGTA503610EA 616 EUR
ORIGTA503610
Anti-ACSS2 Mouse Monoclonal Antibody [clone: OTI2C6]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: ACSS2 (acyl-CoA synthetase short-chain family member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR