Anti-CYP2J2 Mouse Monoclonal Antibody [clone: OTI5C5]

Supplier: OriGene
ORIGTA503636EA 603 EUR
ORIGTA503636
Anti-CYP2J2 Mouse Monoclonal Antibody [clone: OTI5C5]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: CYP2J2 (cytochrome P450, family 2, subfamily J, polypeptide 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR