Anti-RBBP7 Mouse Monoclonal Antibody [clone: OTI5A4]

Supplier: OriGene
TA503667
ORIGTA503667EA 593 EUR
ORIGTA503667
Anti-RBBP7 Mouse Monoclonal Antibody [clone: OTI5A4]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: RBBP7 (retinoblastoma binding protein 7)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR