Anti-DAND5 Mouse Monoclonal Antibody [clone: OTI3D4]

Supplier: OriGene
ORIGTA503675EA 603 EUR
ORIGTA503675
Anti-DAND5 Mouse Monoclonal Antibody [clone: OTI3D4]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: DAND5 (DAN domain family member 5, BMP antagonist)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR