Anti-NIF3L1 Mouse Monoclonal Antibody [clone: OTI5A8]

Supplier: OriGene
ORIGTA503712EA 603 EUR
ORIGTA503712
Anti-NIF3L1 Mouse Monoclonal Antibody [clone: OTI5A8]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: NIF3L1 (NIF3 NGG1 interacting factor 3-like 1 (S. cerevisiae))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR