Anti-PDE6G Mouse Monoclonal Antibody [clone: OTI2F5]

Supplier: OriGene
ORIGTA503736EA 603 EUR
ORIGTA503736
Anti-PDE6G Mouse Monoclonal Antibody [clone: OTI2F5]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: PDE6G (phosphodiesterase 6G, cGMP-specific, rod, gamma)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR