Anti-KATNB1 Mouse Monoclonal Antibody [clone: OTI5H6]

Supplier: OriGene
TA503750
ORIGTA503750EA 616 EUR
ORIGTA503750
Anti-KATNB1 Mouse Monoclonal Antibody [clone: OTI5H6]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: KATNB1 (katanin p80 (WD repeat containing) subunit B 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR