Anti-LPL Mouse Monoclonal Antibody [clone: OTI3A10]

Supplier: OriGene
ORIGTA503793EA 603 EUR
ORIGTA503793
Anti-LPL Mouse Monoclonal Antibody [clone: OTI3A10]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: LPL (lipoprotein lipase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR