Anti-ACSS2 Mouse Monoclonal Antibody [clone: OTI2G8]

Supplier: OriGene
ORIGTA503833EA 603 EUR
ORIGTA503833
Anti-ACSS2 Mouse Monoclonal Antibody [clone: OTI2G8]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: ACSS2 (acyl-CoA synthetase short-chain family member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR