Anti-LDLRAP1 Mouse Monoclonal Antibody [clone: OTI3H10]

Supplier: OriGene
ORIGTA503845EA 603 EUR
ORIGTA503845
Anti-LDLRAP1 Mouse Monoclonal Antibody [clone: OTI3H10]
Antibodies
  • Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LDLRAP1 (low density lipoprotein receptor adaptor protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR