Anti-NAPEPLD Mouse Monoclonal Antibody [clone: OTI7B9]

Supplier: OriGene
TA503861
ORIGTA503861EA 616 EUR
ORIGTA503861
Anti-NAPEPLD Mouse Monoclonal Antibody [clone: OTI7B9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: NAPEPLD (N-acyl phosphatidylethanolamine phospholipase D)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR