Anti-HARS2 Mouse Monoclonal Antibody [clone: OTI6F9]

Supplier: OriGene
ORIGTA503866EA 603 EUR
ORIGTA503866
Anti-HARS2 Mouse Monoclonal Antibody [clone: OTI6F9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: HARS2 (histidyl-tRNA synthetase 2, mitochondrial)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR