Anti-CENPH Mouse Monoclonal Antibody [clone: OTI4G6]

Supplier: OriGene
ORIGTA503870EA 603 EUR
ORIGTA503870
Anti-CENPH Mouse Monoclonal Antibody [clone: OTI4G6]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: CENPH (centromere protein H)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR