Anti-RHOD Mouse Monoclonal Antibody [clone: OTI2F7]

Supplier: OriGene
ORIGTA503891EA 608 EUR
ORIGTA503891
Anti-RHOD Mouse Monoclonal Antibody [clone: OTI2F7]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: RHOD (ras homolog family member D)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR