Anti-PDIA4 Mouse Monoclonal Antibody [clone: OTI2E2]

Supplier: OriGene
ORIGTA503904EA 603 EUR
ORIGTA503904
Anti-PDIA4 Mouse Monoclonal Antibody [clone: OTI2E2]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PDIA4 (protein disulfide isomerase family A, member 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR