Anti-TUBAL3 Mouse Monoclonal Antibody [clone: OTI3B7]

Supplier: OriGene
ORIGTA503932EA 603 EUR
ORIGTA503932
Anti-TUBAL3 Mouse Monoclonal Antibody [clone: OTI3B7]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TUBAL3 (tubulin, alpha-like 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR