Anti-FBXO21 Mouse Monoclonal Antibody [clone: OTI1E1]

Supplier: OriGene
ORIGTA503940EA 581 EUR
ORIGTA503940
Anti-FBXO21 Mouse Monoclonal Antibody [clone: OTI1E1]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: FBXO21 (F-box protein 21)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR