Anti-DPP9 Mouse Monoclonal Antibody [clone: OTI4H8]

Supplier: OriGene
ORIGTA503956EA 603 EUR
ORIGTA503956
Anti-DPP9 Mouse Monoclonal Antibody [clone: OTI4H8]
Antibodies
  • Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: DPP9 (dipeptidyl-peptidase 9)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR