Anti-PDSS2 Mouse Monoclonal Antibody [clone: OTI1C4]

Supplier: OriGene
ORIGTA503975EA 603 EUR
ORIGTA503975
Anti-PDSS2 Mouse Monoclonal Antibody [clone: OTI1C4]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PDSS2 (prenyl (decaprenyl) diphosphate synthase, subunit 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR