Anti-SDS Mouse Monoclonal Antibody [clone: OTI2C11]

Supplier: OriGene
ORIGTA504009EA 586 EUR
ORIGTA504009
Anti-SDS Mouse Monoclonal Antibody [clone: OTI2C11]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SDS (serine dehydratase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR