Anti-TPSG1 Mouse Monoclonal Antibody [clone: OTI3F2]

Supplier: OriGene
ORIGTA504030EA 603 EUR
ORIGTA504030
Anti-TPSG1 Mouse Monoclonal Antibody [clone: OTI3F2]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: TPSG1 (tryptase gamma 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR