Anti-DPP9 Mouse Monoclonal Antibody [clone: OTI6A12]

Supplier: OriGene
ORIGTA504039EA 603 EUR
ORIGTA504039
Anti-DPP9 Mouse Monoclonal Antibody [clone: OTI6A12]
Antibodies
  • Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: DPP9 (dipeptidyl-peptidase 9)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR