Anti-PDIA4 Mouse Monoclonal Antibody [clone: OTI2B9]

Supplier: OriGene
ORIGTA504129EA 603 EUR
ORIGTA504129
Anti-PDIA4 Mouse Monoclonal Antibody [clone: OTI2B9]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: PDIA4 (protein disulfide isomerase family A, member 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR