Anti-TULP3 Mouse Monoclonal Antibody [clone: OTI1A10]

Supplier: OriGene
TA504148
ORIGTA504148EA 598 EUR
ORIGTA504148
Anti-TULP3 Mouse Monoclonal Antibody [clone: OTI1A10]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: TULP3 (tubby like protein 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR