Anti-SSX5 Mouse Monoclonal Antibody [clone: OTI1D6]

Supplier: OriGene
ORIGTA504221EA 581 EUR
ORIGTA504221
Anti-SSX5 Mouse Monoclonal Antibody [clone: OTI1D6]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SSX5 (synovial sarcoma, X breakpoint 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR