Anti-EXOSC7 Mouse Monoclonal Antibody [clone: OTI1G8]

Supplier: OriGene
ORIGTA504239EA 608 EUR
ORIGTA504239
Anti-EXOSC7 Mouse Monoclonal Antibody [clone: OTI1G8]
Antibodies
  • Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: EXOSC7 (exosome component 7)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR