Anti-C1S Mouse Monoclonal Antibody [clone: OTI2D11]

Supplier: OriGene
ORIGTA504246EA 581 EUR
ORIGTA504246
Anti-C1S Mouse Monoclonal Antibody [clone: OTI2D11]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: C1S (complement component 1, s subcomponent)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR