Anti-ALOX15 Mouse Monoclonal Antibody [clone: OTI3G8]

Supplier: OriGene
ORIGTA504250EA 609 EUR
ORIGTA504250
Anti-ALOX15 Mouse Monoclonal Antibody [clone: OTI3G8]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: ALOX15 (arachidonate 15-lipoxygenase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR