Anti-MIER2 Mouse Monoclonal Antibody [clone: OTI3G7]

Supplier: OriGene
ORIGTA504260EA 616 EUR
ORIGTA504260
Anti-MIER2 Mouse Monoclonal Antibody [clone: OTI3G7]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: MIER2 (mesoderm induction early response 1, family member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR