Anti-RFXANK Mouse Monoclonal Antibody [clone: OTI2C6]

Supplier: OriGene
ORIGTA504278EA 603 EUR
ORIGTA504278
Anti-RFXANK Mouse Monoclonal Antibody [clone: OTI2C6]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: RFXANK (regulatory factor X-associated ankyrin-containing protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR