Anti-RGS16 Mouse Monoclonal Antibody [clone: OTI2C9]

Supplier: OriGene
ORIGTA504308EA 603 EUR
ORIGTA504308
Anti-RGS16 Mouse Monoclonal Antibody [clone: OTI2C9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: RGS16 (regulator of G-protein signaling 16)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR