Anti-PPP1R15A Mouse Monoclonal Antibody [clone: OTI4F1]

Supplier: OriGene
ORIGTA504313EA 593 EUR
ORIGTA504313
Anti-PPP1R15A Mouse Monoclonal Antibody [clone: OTI4F1]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PPP1R15A (protein phosphatase 1, regulatory subunit 15A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR