Anti-CST2 Mouse Monoclonal Antibody [clone: OTI1H8]

Supplier: OriGene
TA504331
ORIGTA504331EA 593 EUR
ORIGTA504331
Anti-CST2 Mouse Monoclonal Antibody [clone: OTI1H8]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CST2 (cystatin SA)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR