Anti-CST2 Mouse Monoclonal Antibody [clone: OTI5G8]

Supplier: OriGene
ORIGTA504340EA 581 EUR
ORIGTA504340
Anti-CST2 Mouse Monoclonal Antibody [clone: OTI5G8]
Antibodies
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: CST2 (cystatin SA)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR