Anti-SH2D2A Mouse Monoclonal Antibody [clone: OTI6G1]

Supplier: OriGene
TA504461
ORIGTA504461EA 616 EUR
ORIGTA504461
Anti-SH2D2A Mouse Monoclonal Antibody [clone: OTI6G1]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: SH2D2A (SH2 domain containing 2A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR