Anti-SCHIP1 Mouse Monoclonal Antibody [clone: OTI2E7]

Supplier: OriGene
ORIGTA504468EA 581 EUR
ORIGTA504468
Anti-SCHIP1 Mouse Monoclonal Antibody [clone: OTI2E7]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SCHIP1 (schwannomin interacting protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR