Anti-CNOT4 Mouse Monoclonal Antibody [clone: OTI3A3]

Supplier: OriGene
ORIGTA504472EA 603 EUR
ORIGTA504472
Anti-CNOT4 Mouse Monoclonal Antibody [clone: OTI3A3]
Antibodies
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: CNOT4 (CCR4-NOT transcription complex, subunit 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR