Anti-S100P Mouse Monoclonal Antibody [clone: OTI1F1]

Supplier: OriGene
ORIGTA504477EA 581 EUR
ORIGTA504477
Anti-S100P Mouse Monoclonal Antibody [clone: OTI1F1]
Antibodies
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: S100P (S100 calcium binding protein P)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR