Anti-PPAT Mouse Monoclonal Antibody [clone: OTI1D9]

Supplier: OriGene
TA504508
ORIGTA504508EA 616 EUR
ORIGTA504508
Anti-PPAT Mouse Monoclonal Antibody [clone: OTI1D9]
Recommended Dilutions: Western Blot: 1:200-1:1000

Type: Primary
Antigen: PPAT (phosphoribosyl pyrophosphate amidotransferase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR