Anti-CPA1 Mouse Monoclonal Antibody [clone: OTI4F10]

Supplier: OriGene
TA504522
ORIGTA504522EA 593 EUR
ORIGTA504522
Anti-CPA1 Mouse Monoclonal Antibody [clone: OTI4F10]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CPA1 (carboxypeptidase A1 (pancreatic))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR