Anti-LHX1 Mouse Monoclonal Antibody [clone: OTI1D1]

Supplier: OriGene
ORIGTA504532EA 603 EUR
ORIGTA504532
Anti-LHX1 Mouse Monoclonal Antibody [clone: OTI1D1]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: LHX1 (LIM homeobox 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR