Anti-FOLH1 Mouse Monoclonal Antibody [clone: OTI3E8]

Supplier: OriGene
ORIGTA504550EA 603 EUR
ORIGTA504550
Anti-FOLH1 Mouse Monoclonal Antibody [clone: OTI3E8]
Antibodies
  • Recommended Dilutions: Western Blot: 1:1000-2000

Type: Primary
Antigen: FOLH1 (folate hydrolase (prostate-specific membrane antigen) 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR