Anti-FXYD3 Mouse Monoclonal Antibody [clone: OTI1A8]

Supplier: OriGene
ORIGTA504572EA 603 EUR
ORIGTA504572
Anti-FXYD3 Mouse Monoclonal Antibody [clone: OTI1A8]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: FXYD3 (FXYD domain containing ion transport regulator 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR